Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP3030

Sigma-Aldrich

ENA-78 (8-78) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CXCL5, Epithelial Neutrophil Activating Peptide-78, LIX (murine)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

10.0-100.0 ng/mL

mol wt

7.8 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CXCL5(6374)

General description

ENA-78 (epithelial cell-derived neutrophil activating peptide-78) is a CXC chemokine that signals through the CXCR2 receptor. It is expressed in monocytes, platelets, endothelial cells, and mast cells. ENA-78 is a chemoattractant for neutrophils. The murine homolog of ENA-78 is called LIX. Three N-terminal truncated variants of human ENA-78; ENA 5-78, ENA 8-78, ENA 9-78, contain 74, 71, and 70 amino acid residues, respectively, possess increased biological activity. ENA-78 contains the four conserved cysteine residues present in CXC chemokines, and also contains the ‘ELR′ motif common to CXC chemokine that bind to the CXCR1 and CXCR2 receptors. Recombinant human ENA-78 is a 7.8 kDa protein consisting of 71 amino acid residues. The ENA-78 gene is mapped to human chromosome 4q13.3.

Biochem/physiol Actions

ENA-78 (epithelial cell-derived neutrophil activating peptide-78) gene expression is associated with sepsis, atherosclerosis, COPD (chronic obstructive pulmonary disease), psoriasis and gastrointestinal inflammation. ENA-78 is angiogenic factor and is associated with cancer progression. Upregulation of ENA-78 gene is observed in non‐small‐cell lung cancer, head, neck, gastric, pancreatic, colorectal, prostate and lung cancer squamous cell carcinoma.
ENA-78 is a CXC chemokine that signals through the CXCR2 receptor. It is expressed in monocytes, platelets, endothelial cells, and mast cells. Recombinant human ENA-78 is a 7.8 kDa protein consisting of 71 amino acid residues.

Sequence

LRELRCVCLQ TTQGVHPKMI SNLQVFAIGP QCSKVEVVAS LKNGKEICLD PEAPFLKKVI QKILDGGNKE N

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CXCL5 (chemokine (CXC motif) ligand 5).
Bulysheva A A and Yeudall W A
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2013)
Interleukin?17 regulates expression of the CXC chemokine LIX/CXCL5 in osteoblasts: implications for inflammation and neutrophil recruitment.
Ruddy M J, et al.
Journal of Leukocyte Biology, 76(1), 135-144 (2004)
Chemokine receptors in inflammation: an overview.
D'Ambrosio D, et al.
Journal of Immunological Methods, 273(1-2), 3-13 (2003)
CXC chemokines in angiogenesis.
Belperio J A, et al.
Journal of Leukocyte Biology, 68(1), 1-8 (2000)
CXC chemokines in angiogenesis of cancer.
Strieter R M, et al.
Seminars in Cancer Biology, 14(3), 195-200 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service