Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3020

Sigma-Aldrich

sCD22 human

recombinant, expressed in CHO cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

B-Lymphocyte cell adhesion molecule, BL-CAM, Leu-14, Siglec-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in CHO cells

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

10-17 μg/mL ED50

mol wt

75.0 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CD22(933)

General description

CD22 is a B-lineage restricted 135 kDa glycoprotein whose cell surface expression is limited to resting and activated B lymphocytes. The physiological role of CD22 is still unknown. Targeted disruption of CD22 in mice results in a reduced level of surface IgM on peripheral B cells suggesting a role for CD22 in limiting antigen receptor signaling. CD22 is a member of the Ig gene superfamily that uniquely binds a sialic acid-dependent ligand. Recombinant Human sCD22 is a soluble 75.0 kDa protein which corresponds to the extracellular domain of CD22.

Biochem/physiol Actions

CD22 is a B-lineage restricted 135 kDa glycoprotein whose cell surface expression is limited to resting and activated B lymphocytes. Recombinant Human sCD22 is a soluble 75.0 kDa protein which corresponds to the extracellular domain of CD22.

Sequence

SKWVFEHPET LYAWEGACVW IPCTYRALDG DLESFILFHN PEYNKNTSKF DGTRLYESTK DGKVPSEQKR VQFLGDKNKN CTLSIHPVHL NDSGQLGLRM ESKTEKWMER IHLNVSERPF PPHIQLPPEI QESQEVTLTC LLNFSCYGYP IQLQWLLEGV PMRQAAVTST SLTIKSVFTR SELKFSPQWS HHGKIVTCQL QDADGKFLSN DTVQLNVKHT PKLEIKVTPS DAIVREGDSV TMTCEVSSSN PEYTTVSWLK DGTSLKKQNT FTLNLREVTK DQSGKYCCQV SNDVGPGRSE EVFLQVQYAP EPSTVQILHS PAVEGSQVEF LCMSLANPLP TNYTWYHNGK EMQGRTEEKV HIPKILPWHA GTYSCVAENI LGTGQRGPGA ELDVQYPPKK VTTVIQNPMP IREGDTVTLS CNYNSSNPSV TRYEWKPHGA WEEPSLGVLK IQNVGWDNTT IACARCNSWC SWASPVALNV QYAPRDVRVR KIKPLSEIHS GNSVSLQCDF SSSHPKEVQF FWEKNGRLLG KESQLNFDSI SPEDAGSYSC WVNNSIGQTA SKAWTLEVLY APRRLRVSMS PGDQVMEGKS ATLTCESDAN PPVSHYTWFD WNNQSLPHHS QKLRLEPVKV QHSGAYWCQG TNSVGKGRSP LSTLTVYYSP ETIGRR

Physical form

Lyophilized from 1x PBS, pH 7.2.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Cesano et al.
Seminars in oncology, 30(2), 253-257 (2003-04-30)
CD22 is a 135-kd B-cell restricted sialoglycoprotein present in the cytoplasm of virtually all B-lineage cells but expressed on the B-cell surface only at mature stages of differentiation. In humans, the vast majority of IgM(+)IgD(+) B cells express cell-surface CD22

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service