Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2084

Sigma-Aldrich

VHL human

recombinant, expressed in insect cells, ≥70% (SDS-PAGE)

Synonym(s):

HRCA1, RCA1, VHL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

recombinant

expressed in insect cells

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~26 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

650 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... VHL(7428)

Biochem/physiol Actions

von Hippel-Lindau (VHL) disease is a hereditary cancer with a predilection for the central nervous system and retina. The von Hippel-Lindau tumor suppressor gene is mutated in families with von Hippel-Lindau disease and encodes a protein (VHL) of 213 amino acids with an acidic pentapeptide motif in the N-terminus. Mutations in the VHL gene result in constitutive expression of many hypoxia-induced genes, at least in part because of increases in the cellular level of hypoxia-inducible transcription factor HIF-1a. VHL protein binds to elongin B, elongin C, and Cul2 to form a stable complex that targets hypoxia inducible factors (HIFs) for degradation and transcriptional regulation. In addition, VHL protein has also been shown to interact with specific protein kinase C isoforms, histone deacetylases and HIF-1 inhibitor (HIF-1).

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linehan, W.M., et al.
Journal of the American Medical Association, 273, 564-570 (1995)
A D Singh et al.
Survey of ophthalmology, 46(2), 117-142 (2001-10-02)
In recent years advances have been made in the clinical and genetic aspects of von Hippel-Lindau disease (VHL). Retinal capillary hemangioma is the most common manifestation of VHL disease and, therefore, ophthalmologists are frequently involved in the care of patients
F Latif et al.
Science (New York, N.Y.), 260(5112), 1317-1320 (1993-05-28)
A gene discovered by positional cloning has been identified as the von Hippel-Lindau (VHL) disease tumor suppressor gene. A restriction fragment encompassing the gene showed rearrangements in 28 of 221 VHL kindreds. Eighteen of these rearrangements were due to deletions
E Hasanov et al.
Oncogene, 36(24), 3450-3463 (2017-01-24)
The hypoxia-regulated tumor-suppressor von Hippel-Lindau (VHL) is an E3 ligase that recognizes its substrates as part of an oxygen-dependent prolyl hydroxylase (PHD) reaction, with hypoxia-inducible factor α (HIFα) being its most notable substrate. Here we report that VHL has an
Jung Jin Kim et al.
Genes & development, 29(21), 2244-2257 (2015-11-08)
The von Hippel-Lindau tumor suppressor pVHL is an E3 ligase that targets hypoxia-inducible factors (HIFs). Mutation of VHL results in HIF up-regulation and contributes to processes related to tumor progression such as invasion, metastasis, and angiogenesis. However, very little is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service