Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0695

Sigma-Aldrich

TAU-441, TTBK1-Phosphorylated

recombinant

Synonym(s):

Phosphorylated TAU antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in E. coli

Assay

70-95% (SDS-PAGE)

form

frozen liquid

mol wt

~64-70 kDa

concentration

0.2 μg/μL

accession no.

P10636-8

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAPT(4137)

Application

Useful for kinase assay and Western blot.

Biochem/physiol Actions

Recombinant human tag-free Tau-441 was co-expressed with TTBK1 in E. coli cells. The Tau-441 was phosphorylated by TTBK1 in vivo and in vitro prior to the final chromatography purification. Tau-441 or Tau-F is a member of the Tau family of proteins which function to stabilize the microtubules by binding to them. Tau proteins are subject to phosphorylation and this phenomenon regulates the association of the Tau protein with the microtubules (1). Deposits of Alzheimer′s disease AD-associated proteins, such as hyperphosphorylated Tau, as well as other shared misfolded proteins, such as, β-amyloid precursor protein (βAPP), ubiquitin, and various chaperones and protein kinases are thought to play a pathologic role in the cognitive decline and muscular failure. Malfunctioning of Tau proteins is associated with microtubules disintegration and collapsing of the neuronal transport system (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service