Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP0527

Sigma-Aldrich

Mouse PD-1 (CD279), FC Fusion

recombinant, expressed in HEK 293 cells

Synonym(s):

CD279, PDCD1, Programmed Cell Death 1, SLEB2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

43.2 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... PDCD1(18566)

General description

Mouse secreted Programmed Cell Death 1 (PD-1)-Fc fusion protein, also known as CD279, PDCD1, and SLEB2, GenBank Accession No. NM_008798, a.a. 25-167,fused at the C-terminus to the Fc portion of mouse IgG2a, expressed in a HEK293 cell expression system. MW = 43.2 kDa. Note:this protein runs at an apparent higher molecular weight due to glycosylation.

Physical form

Aqueous solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service