Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP0360

Sigma-Aldrich

PLK1 Polo Box Domain active human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

STPK13, polo-like kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

53 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

2.9 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PLK1(5347)

General description

Human Plk1 polo box domain (GenBank Accession No. NM_005030), amino acids 367-end with N-terminal GST-tag, MW=53 kDa, expressed in an E. coli expression system.
Polo-like kinase 1 (PLK1) belongs to the PLK family. It comprises polo-box domain (PBD). PBD, in turn, contains polo boxes (PB1 and PB2) with a 6-stranded β sandwich and an α helix as structural elements. The PLK1 gene is mapped to human chromosome 16p12.2.

Application

PLK1 Polo Box Domain active human has been used in the in vitro binding assay with various synthetic peptides coupled to streptavidin beads. It has also been used in binding studies with fluorescein-labeled substrate peptide 5-carboxyfluorescein-GPMQSpTPLNG to test the effect of various inhibitors.

Biochem/physiol Actions

Polo-like kinase 1 (PLK1) is a mitotic kinase that mediates the regulation of various events in mitosis including chromosome segregation, cytokinesis, centrosome maturation, and separation. It activates cell division cycle 25 (CDC25) phosphatase which further regulates mitotic entry. Elevated expression of PLK1 is observed in many tumors.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sugunadevi Sakkiah et al.
PloS one, 9(7), e101405-e101405 (2014-07-19)
The polo-like kinase 1 (Plk1) is a critical regulator of cell division that is overexpressed in many types of tumors. Thus, a strategy in the treatment of cancer has been to target the kinase activity (ATPase domain) or substrate-binding domain
Sara Abdelfatah et al.
Journal of advanced research, 16, 145-156 (2019-03-23)
Polo-like kinase 1 (PLK1), a member of the Polo-like kinase family, plays an important regulatory role in mitosis and cell cycle progression. PLK1 overexpression is correlated with tumourigenesis and poor prognosis in cancer patients. Therefore, the identification of novel compounds
Zsofia A Novak et al.
Developmental cell, 37(6), 545-557 (2016-06-22)
Centrosomes and cilia are organized by a centriole pair comprising an older mother and a younger daughter. Centriole numbers are tightly regulated, and daughter centrioles (which assemble in S phase) cannot themselves duplicate or organize centrosomes until they have passed
S-L Zhou et al.
European review for medical and pharmacological sciences, 23(5), 2084-2091 (2019-03-28)
Our study aimed to explore the effects of miRNA-296-5p on the biological behaviors of papillary thyroid carcinoma (PTC) cells and its potential mechanism. Twenty-eight PTC tissues and the corresponding non-cancerous tissues were collected. Real Time-quantitative Polymerase Chain Reaction (RT-qPCR) analysis
Pooja Sharma et al.
Scientific reports, 9(1), 15930-15930 (2019-11-07)
The human polo-like kinase PLK1 coordinates mitotic chromosome segregation by phosphorylating multiple chromatin- and kinetochore-binding proteins. How PLK1 activity is directed to specific substrates via phosphopeptide recognition by its carboxyl-terminal polo-box domain (PBD) is poorly understood. Here, we combine molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service