Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0344

Sigma-Aldrich

EGFR active mouse

recombinant, expressed in baculovirus infected Sf9 cells

Synonym(s):

ERBB, ERRP, Wa5, epidermal growth factor receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in baculovirus infected Sf9 cells

form

aqueous solution

mol wt

62 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... EGFR(13649)

General description

Mouse EGFR (GenBank Accession No. NM_207655/ NP_997538), amino acids 670-1210 with C-terminal Flag-tag, MW = 62 kDa, expressed in SF9 cells via a Baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ariel Talavera et al.
Molecular immunology, 48(12-13), 1578-1585 (2011-05-20)
The EGF receptor is an important target of cancer immunotherapies. The 7A7 monoclonal antibody has been raised against the murine EGFR, but it cross-reacts with the human receptor. The results from experiments using immune-competent mice can therefore, in principle, be
Jeffrey D Kearns et al.
Molecular cancer therapeutics, 14(7), 1625-1636 (2015-04-26)
Although EGFR is a validated therapeutic target across multiple cancer indications, the often modest clinical responses to current anti-EGFR agents suggest the need for improved therapeutics. Here, we demonstrate that signal amplification driven by high-affinity EGFR ligands limits the capacity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service