Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0288

Sigma-Aldrich

ATP Citrate Lyase Active human

recombinant, expressed in baculovirus, ≥90% (SDS-PAGE)

Synonym(s):

ACLY, ATP-citrate synthase, ATPCL, CLATP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

recombinant

expressed in baculovirus

Assay

≥90% (SDS-PAGE)

form

aqueous solution

specific activity

≥1.2 pmol/min-μg

mol wt

147 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... ACLY(47)

General description

Human ATP citrate lyase (ACLY) (GenBank Accession No. NM_001096), full length, with N-terminal GST tag, MW = 147 kDa, expressed in baculovirus expression system.

Application

Active human ATP citrate lyase is useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling. Active human ATP citrate lyase has been used in a study to ascertain the nature of the catalytic phosphorylation that initiates the ACL reaction, and to identity the active site residues involved. [1] Active human ATP citrate lyase has also been used in a study to analyze tumor metabolism to reveal mitochondrial glucose oxidation in genetically diverse human glioblastomas. [2]

Biochem/physiol Actions

ATP citrate lyase is an enzyme involved in fatty acid synthesis that generates cytosolic acetyl-CoA and oxaloacetate from citrate and CoA. ATP citrate lyase is often upregulated in cancer.

Unit Definition

One unit is defined as the amount of enzyme required to convert 1 pmol of ADP to ATP/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20 and 10% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabiola Márquez et al.
Critical reviews in food science and nutrition, 52(7), 585-594 (2012-04-26)
Several studies have shown that Garcinia cambogia plays an important role in the regulation of endogenous lipid biosynthesis. This effect is specially attributed to (-)-hydroxycitric acid (HCA) inhibiting the enzyme ATP-dependent citrate lyase, which catalyzes the cleavage of citrate to
Isaac Marin-Valencia et al.
Cell metabolism, 15(6), 827-837 (2012-06-12)
Dysregulated metabolism is a hallmark of cancer cell lines, but little is known about the fate of glucose and other nutrients in tumors growing in their native microenvironment. To study tumor metabolism in vivo, we used an orthotopic mouse model
Fan Fan et al.
Biochemistry, 51(25), 5198-5211 (2012-06-05)
ATP citrate lyase (ACL) catalyzes an ATP-dependent biosynthetic reaction which produces acetyl-coenzyme A and oxaloacetate from citrate and coenzyme A (CoA). Studies were performed with recombinant human ACL to ascertain the nature of the catalytic phosphorylation that initiates the ACL

Articles

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service