Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0235

Sigma-Aldrich

PI3 kinase [p110a(H1047R)/p85a] Active human

recombinant, expressed in baculovirus infected insect cells, ≥60% (SDS-PAGE)

Synonym(s):

PIK3CA, Phosphoinositide-3-kinase, catalytic α, PtdIns-3-kinase p110

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥60% (SDS-PAGE)

form

aqueous solution

specific activity

≥1770 pmol/min-μg

mol wt

129 kDa (p110α)
88 kDa (p85α)

packaging

pkg of 20 μg

concentration

0.38 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PIK3CA(5290)

General description

Complex of N-terminal FLAG-tagged recombinant full-length human p110α (GenBank Accession No. U79143), with H1047R mutation, and recombinant full length, human p85α (no tag, GenBank Accession No. XM_043865). Co-expressed in Sf9 cells in a Baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit of kinase activity is defined as the amount of enzyme required to produce 1 pmol of Phosphoinositol-3, 4,5-Trisphosphate/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 30% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service