Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRE0053

Sigma-Aldrich

Lysostaphin from Staphylococcus staphylolyticus

Suitable for manufacturing of diagnostic kits and reagents., lyophilized powder

Synonym(s):

Glycyl-glycine Endopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

bacterial (Staphylococcus sp.)

Quality Level

form

lyophilized powder

specific activity

≥500 units/mg protein

mol wt

26,926 Da

composition

protein, 50-70% biuret

packaging

pkg of (package size sold on basis of mg protein)

concentration

50—70% protein

suitability

suitable for microbiological tests

application(s)

diagnostic assay manufacturing

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Lysostaphin is a glycylglycine endopeptidase. It is an important antibacterial enzyme.[1]

Biochem/physiol Actions

Lysostaphin has the ability to cleave both viable and heat-killed S. aureus cells. It is one of the major antibacterial agents for the staphylococci.[2] Lysostaphin cleaves the cross-linking pentaglycine bridges in the cell walls of staphylococci. Thus it is considered as an antistaphylococcal therapeutic agent.[1]
Lysostaphin is a zinc endopeptidase with a molecular weight of approximately 25 kDa. Because lysostaphin cleaves the polyglycine cross-links in the peptidoglycan layer of the cell wall of Staphylococcus species it has been found useful for cell lysis and also as a potential anti-microbial therapeutic.
pH Optimum for activity: ~7.5

Unit Definition

One unit will reduce the turbidity (A620) of a suspension of Staphylococcus aureus cells from 0.250 to 0.125 in 10 min at pH7.5 at 37°C in a 6.0ml reaction mixture.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysostaphin: a new bacteriolytic agent for the Staphylococcus
Schindler, Ch A and Schuhardt, VT
Proceedings of the National Academy of Sciences of the USA, 51(3), 414-421 (1964)
Lysostaphin disrupts Staphylococcus aureus and Staphylococcus epidermidis biofilms on artificial surfaces
Wu J A, et al.
Antimicrobial Agents and Chemotherapy, 47(11), 3407-3414 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service