Skip to Content
MilliporeSigma

SML3976

Apremilast

≥98% (HPLC)

Synonym(s):

(S)-N-{2-[1-(3-Ethoxy-4-methoxyphenyl)-2-methanesulphonylethyl]-1,3-dioxo-2,3-dihydro-1H-isoindol-4-yl}acetamide, CC 10004, CC-10004, CC10004, N-[2-[(1S)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl]-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H24N2O7S
CAS Number:
Molecular Weight:
460.50
UNSPSC Code:
12352200
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

[S](=O)(=O)(C[C@@H](N2C(=O)c3c(cccc3NC(=O)C)C2=O)c1cc(c(cc1)OC)OCC)C

InChI

1S/C22H24N2O7S/c1-5-31-19-11-14(9-10-18(19)30-3)17(12-32(4,28)29)24-21(26)15-7-6-8-16(23-13(2)25)20(15)22(24)27/h6-11,17H,5,12H2,1-4H3,(H,23,25)/t17-/m1/s1

InChI key

IMOZEMNVLZVGJZ-QGZVFWFLSA-N

Biochem/physiol Actions

Orally active, potent and selective phosphodiesterase 4 (PDE4) inhibitor in vitro and in vivo.



Apremilast (CC-10004) is an orally active, potent and selective phosphodiesterase 4 (PDE4) inhibitor (IC50 = 20/49/50/30 nM with recombinant human PDE4A4/B2/C2/D3 and 1 μM cAMP; no significant PDE1/2/3/5/7/8/9/10/11 inhibition at 10 μM). Apremilast effectively inhibits LPS-induced cytokine and chemokine protein expression from purified human PBMCs (TNFα/IFNγ/IL12 IC50 = 13/110/120 nM, IP10/MIG/MIP1α IC50 = 10/28/40 nM) in vitro and exhibits therapeutic efficacy in a psoriasis mouse model in vivo (2.5/mg/kg b.i.d. p.o.)


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - STOT SE 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library