Skip to Content
MilliporeSigma

SML3965

Procyanidin B2

≥97% (HPLC), anti-inflammatory, powder

Synonym(s):

(+)-Procyanidin B2, (-)-Epicatechin-(4β→8)-(-)-epicatechin, (2R,2′R,3R,3′R,4R)-2,2′-Bis(3,4-dihydroxyphenyl)-3,3′,4,4′-tetrahydro[4,8′-bi-2H-1-benzopyran]-3,3′,5,5′,7,7′-hexol, Proanthocyanidin B2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H26O12
CAS Number:
Molecular Weight:
578.52
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥97% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Procyanidin B2, ≥97% (HPLC)

Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to pink

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O1[C@@H]([C@@H]([C@H](c6c1cc(cc6O)O)c3c4c(c(cc3O)O)C[C@H]([C@H](O4)c5cc(c(cc5)O)O)O)O)c2cc(c(cc2)O)O

InChI

1S/C30H26O12/c31-13-7-20(37)24-23(8-13)41-29(12-2-4-16(33)19(36)6-12)27(40)26(24)25-21(38)10-17(34)14-9-22(39)28(42-30(14)25)11-1-3-15(32)18(35)5-11/h1-8,10,22,26-29,31-40H,9H2/t22-,26-,27-,28-,29-/m1/s1

InChI key

XFZJEEAOWLFHDH-NFJBMHMQSA-N

Biochem/physiol Actions

Polyphenolic plant compound that exhibits anti-inflammatory, anti-cancer, antiviral, and antioxidant properties.

Procyanidin B2 is a polyphenolic plant compound that exhibits anti-inflammatory, anti-cancer, antiviral, and antioxidant properties. Procyanidin B2 attenuates dyslipidemia, hyperglycemia, and oxidative stress in individuals with metabolic syndrome. Apparently, it reduces fatty liver by remodeling intestinal flora in a high-fat-cholesterol diet fed rabbits.


Still not finding the right product?

Explore all of our products under Procyanidin B2


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library