Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3908

Sigma-Aldrich

SLU-PP-332

≥98% (HPLC)

Synonym(s):

(E)-4-Hydroxy-N′(naphthalen-2-ylmethylene)benzohydrazide, 4-Hydroxybenzoic acid 2-(2-naphthalenylmethylene)hydrazide, ERR pan-Agonist 332, SR9861

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N2O2
CAS Number:
Molecular Weight:
290.32
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

OC1=CC=C(C=C1)C(N/N=C/C2=CC3=C(C=C2)C=CC=C3)=O

InChI key

RNZIMBFHRXYRLL-XDHOZWIPSA-N

Biochem/physiol Actions

Non-toxic pan ERRα/β/γ agonist with significant ERRα activity that acts as an exercise mimetic.



SLU-PP-332 is a non-toxic pan ERRα/β/γ (estrogen receptor-related receptors α/β/γ) agonist with significant ERRα activity (EC50 = 98, 230 & 430 nM against ERRα, ERRβ & ERγ) and acts as an exercise mimetic. SLU-PP-332 displays selectivity over ERα, ERβ and several other closely related nuclear receptors. It induces Ddit4 expression and enhances mitochondrial respiration in C2C12 myocytes (10 µM) and exercise endurance in mice (50 mg/kg bid, ip) in an ERRα-dependent manner.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service