Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SML3852

Sigma-Aldrich

Saroglitazar Magnesium

≥98% (HPLC)

Synonym(s):

(αS)-α-Ethoxy-4-[2-[2-methyl-5-[4-(methylthio)phenyl]-1H-pyrrol-1-yl]ethoxy]benzenepropanoic acid Magnesium, (s)-2-Ethoxy-3-(4-(2-(2-methyl-5-(4-(methylthio)phenyl)-1H-pyrrol-1-yl)ethoxy)phenyl)propanoic acid Magnesium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H56MgN2O8S2
CAS Number:
Molecular Weight:
901.42
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

[Mg+2].S(C)c4ccc(cc4)c5[n](c(cc5)C)CCOc6ccc(cc6)C[C@H](OCC)C(=O)[O-].S(C)c1ccc(cc1)c2[n](c(cc2)C)CCOc3ccc(cc3)C[C@H](OCC)C(=O)[O-]

InChI

1S/2C25H29NO4S.Mg/c2*1-4-29-24(25(27)28)17-19-6-10-21(11-7-19)30-16-15-26-18(2)5-14-23(26)20-8-12-22(31-3)13-9-20;/h2*5-14,24H,4,15-17H2,1-3H3,(H,27,28);/q;;+2/p-2/t2*24-;/m00./s1

InChI key

UJYFZCVPOSZDMK-YPPDDXJESA-L

Biochem/physiol Actions

Potent and selective PPARα/γ agonist.

Saroglitazar is a potent and selective PPARα/γ (peroxisome proliferator-activated receptor) agonist that exhibits lipid-lowering and insulin-sensitizing effects. Saroglitazar is approved in India for the treatment of diabetes type 2 and dyslipidemia.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saroglitazar, a novel dual PPAR-?/? agonist, reverses high fat-low protein diet-induced metabolic and cognitive aberrations in C57BL/6J male mice
Life Sciences, 271, 119191-119191 (2021)
Saroglitazar reduces obesity and associated inflammatory consequences in murine adipose tissue
European Journal of Pharmacology, 822, 32-42 (2018)
Saroglitazar, a novel PPAR?/? agonist with predominant PPAR? activity, shows lipid-lowering and insulin-sensitizing effects in preclinical models
Pharmacology Research & Perspectives, 3(3) (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service