Skip to Content

SML3759

MU1742

≥95% (HPLC)

Synonym(s):

4-(1-((4-Fluoro-1-methylpiperidin-4-yl)methyl)-4-(5-fluoropyridin-2-yl)-1H-imidazol-5-yl)-1H-pyrrolo[2,3-b]pyridine, MU 1742, MU-1742

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H22F2N6
Molecular Weight:
408.45
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC5(CCN(CC5)C)C[n]1cnc(c1c3c4c([nH]cc4)ncc3)c2ncc(cc2)F

InChI key

SWOIFXHMBKFCRM-UHFFFAOYSA-N

Biochem/physiol Actions

MU1742 is an orally active, potent and selective casein kinase 1 inhibitor (CK1α/δ/ε IC50 = 7/6/28 nM; p38α IC50 >3 μM) with high kinome-wide selectivity. MU1742 inhibits CLL19-induced chemotaxis (IC50 = 200 nM; MEC1 cells) and CK1δ/ε-dependent DVL3 phosphorylation (IC50 = 200 nM post 18h treatment; HL60 cells) in cultures and in lung tissue in mice in vivo (5h post 100 mg/kg p.o. using HCl salt form).
Orally active, potent and selective casein kinase 1 (CK1, CKI) inhibitor in vitro and in vivo.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library