Skip to Content
MilliporeSigma

SML3750

SR-302

≥98% (HPLC)

Synonym(s):

N-(4-(((S)-4-Cyclohexyl-1-(((SR)-1-(methylsulfonyl)piperidin-3-yl)amino)-1-oxobutan-2-yl)carbamoyl)benzyl)imidazo[1,2-a]-pyridine-3-carboxamide, N-(4-((2S)-4-Cyclohexyl-1-(1-(methylsulfonyl)piperidin-3-ylamino)-1-oxobutan-2-ylcarbamoyl)benzyl)imidazo[1,2-a]pyridine-3-carboxamide, SR 302, SR302

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H42N6O5S
CAS Number:
Molecular Weight:
622.78
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(NCC1=CC=C(C=C1)C(N[C@H](C(NC2CN(CCC2)S(=O)(C)=O)=O)CCC3CCCCC3)=O)C4=CN=C5C=CC=CN54

InChI

1S/C32H42N6O5S/c1-44(42,43)37-18-7-10-26(22-37)35-31(40)27(17-14-23-8-3-2-4-9-23)36-30(39)25-15-12-24(13-16-25)20-34-32(41)28-21-33-29-11-5-6-19-38(28)29/h5-6,11-13,15-16,19,21,23,26-27H,2-4,7-10,14,17-18,20,22H2,1H3,(H,34,41)(H,35,40)(H,36,39)/t26?,27-/m

InChI key

WGMNFJAFXXXRLZ-GEVKEYJPSA-N

Biochem/physiol Actions

Potent ATP-competitive inhibitor against DDR1, DDR2; MAPK11 (p38β), and MAPK14 (p38α) with good cellular target actviity and kinome selectivity.
SR-302 is an ATP-competitive, potent and selective inhibitor against DDR1/2 & MAPK11/14 (IC50/[ATP] = 53.3 nM/100 μM/DDR1, 0.75 nM/5 μM/DDR2; 45.1 nM/50 μM/MAPK11 (p38β), 6.2 nM/2.5 μM/MAPK14 (p38α) by cell-free kinase assay) with good cellular target actviity (IC50 = 18 nM/DDR2, 23 nM/DDR1, 125 nM/MAPK14, 196 nM/MAPK11 by NanoBRET) and kinome selectivity.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library