Skip to Content
MilliporeSigma

SML3638

Ceftobiprole

≥95% (HPLC), Cell wall synthesis inhibitor, powder

Synonym(s):

(6R,7R)-7-[[(2Z)-2-(5-amino-1,2,4-thiadiazol-3-ylidene)- 2-nitroso-1-oxoethyl]amino]-8-oxo-3-[(E)-[2-oxo-1-[(3R)- 3-pyrrolidinyl]-3-pyrrolidinylidene]methyl]-5-thia-1- azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, BAL 9141, BAL 9141-000, Ro 63-9141

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H22N8O6S2
CAS Number:
Molecular Weight:
534.57
UNSPSC Code:
51111800
NACRES:
NA.25
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ceftobiprole, ≥95% (HPLC)

Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Acidified)

storage temp.

-10 to -25°C

SMILES string

[s]1nc(nc1N)C(N=O)C(=O)N[C@H]2[C@H]3SCC(=C(N3C2=O)C(=O)O)\C=C4/CCN(C/4=O)[C@H]5CNCC5

InChI

1S/C20H22N8O6S2/c21-20-24-14(26-36-20)11(25-34)15(29)23-12-17(31)28-13(19(32)33)9(7-35-18(12)28)5-8-2-4-27(16(8)30)10-1-3-22-6-10/h5,10-12,18,22H,1-4,6-7H2,(H,23,29)(H,32,33)(H2,21,24,26)/b8-5+/t10-,11?,12-,18-/m1/s1

InChI key

XLHWFEHWWUCKOL-ADPMQJGOSA-N

Biochem/physiol Actions

Broad-spectrum cephalosporin active against MRSA
Ceftobiprole is a fifth generation broad spectrum cephalosporin that is active against many resistant bacterial strains including methicillin-resistant Staphylococcus aureus (MRSA). Ceftobiprole inhabits PBP1 (penicillin-binding protein 1), PBP2 and ß-lactam resistance determinant PBP2a.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library