Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3542

Sigma-Aldrich

SGC-UBD253

≥98% (HPLC)

Synonym(s):

3-(8-Chloro-3-(2-(2-methoxybenzylamino)-2-oxoethyl)-4-oxo-3,4-dihydroquinazolin-2-yl)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20ClN3O5
Molecular Weight:
429.85
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=C(NCC(C=CC=C1)=C1OC)CN(C(CCC(O)=O)=N2)C(C3=C2C(Cl)=CC=C3)=O

Biochem/physiol Actions

SGC-UBD253 is a cell penetrant, potent and selective chemical probe for the ubiquitin binding domain (UBD) of HDAC6 (histone deacetylase 6). SGC-UBD253 potently binds to HDAC6 UBD (KD = 84 nM) and inhibits the HDAC6-ISG15 interaction (EC50 = 1.9 µM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renato Ferreira de Freitas et al.
Journal of medicinal chemistry, 61(10), 4517-4527 (2018-05-10)
HDAC6 plays a central role in the recruitment of protein aggregates for lysosomal degradation and is a promising target for combination therapy with proteasome inhibitors in multiple myeloma. Pharmacologically displacing ubiquitin from the zinc-finger ubiquitin-binding domain (ZnF-UBD) of HDAC6 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service