Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3534

Sigma-Aldrich

Laninamivir octanoate

≥98% (HPLC)

Synonym(s):

3-(O)-octanoyl R 125489, 3-(O)-octanoyl R-125489, 3-(O)-octanoyl R125489, 4S,5R,6R)-5-Acetamido-4-guanidino-6-((1R,2R)-2-hydroxy-1-methoxy-3-(octanoyloxy)propyl)-5,6-dihydro-4H-pyran-2-carboxylic acid, 5-(Acetylamino)-4-[(aminoiminomethyl)amino]-2,6-anhydro-3,4,5-trideoxy-7-O-methyl-D-glycero-D-galacto-non-2-enonic acid 9-octanoate, CS 8958, CS-8958, CS8958, LNO, Laninamivir prodrug

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H36N4O8
CAS Number:
Molecular Weight:
472.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

CCCCCCCC(OC[C@@H](O)[C@@H](OC)[C@@]1([H])[C@H](NC(C)=O)[C@@H](NC(N)=N)C=C(C(O)=O)O1)=O

InChI

1S/C21H36N4O8/c1-4-5-6-7-8-9-16(28)32-11-14(27)18(31-3)19-17(24-12(2)26)13(25-21(22)23)10-15(33-19)20(29)30/h10,13-14,17-19,27H,4-9,11H2,1-3H3,(H,24,26)(H,29,30)(H4,22,23,25)/t13-,14+,17+,18+,19+/m0/s1

InChI key

UKTIJASCFRNWCB-RMIBSVFLSA-N

Biochem/physiol Actions

Laninamivir (R-125489) prodrug with improved in vivo efficacy than R-125489 and Zanamivir in a mouse influenza virus A (IAV) infection model.
Laninamivir octanoate (CS-8958) is the prodrug form of Laninamivir (R-125489), a potent inhibitor agianst the neuraminidase (NA) activities of various type A and B influenza viruses, including subtypes N1 to N9 and Oseltamivir-resistant strains. When administered intranasally (3X 0.2 μmol/kg; 4 h before, 4 h & 18 h post infection), CS-8958 shows improved in vivo efficacy (100% survival rate on day 20 post infecfion) than R-125489 (100/30/20% survival rate on day 7/10/20 post infecfion) and Zanamivir (100%/0% death on day 7/10) in a mouse influenza virus A/Puerto Rico/8/34 infection model, with prolonged survival effect post a single dosage given even 7 days before infection. Note: Laninamivir octanoate is mainly used for in vivo studies, it exhibits only weak to little antiviral potency before it is converted to the active drug form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service