Skip to Content
MilliporeSigma

SML3490

FMC

≥98% (HPLC)

Synonym(s):

Dimethyl(fluoromethyl)(2-hydroxyethyl)ammonium chloride, Fluorocholine chloride, Fluoromethylcholine chloride, N-(Fluoromethyl)-2-hydroxy-n,n-dimethylethanaminium chloride, NSC720948

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H13FNO·Cl
CAS Number:
Molecular Weight:
157.61
UNSPSC Code:
41106300
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
(Crystals to semi-solid)
Storage condition:
under inert gas
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

(Crystals to semi-solid)

storage condition

under inert gas

color

colorless to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

FC[N+](CCO)(C)C.[Cl-]

InChI

1S/C5H13FNO.ClH/c1-7(2,5-6)3-4-8;/h8H,3-5H2,1-2H3;1H/q+1;/p-1

InChI key

QJEHIIJVWXGJAB-UHFFFAOYSA-M

Biochem/physiol Actions

FMC is a monofluorinated choline that acts as a nonlethal, mechanism-based irreversible inhibtor (pseudosubstrate-based suicide inhibitor) against gut anaerobic bacterial glycyl radical enzyme (GRE) cutC choline trimethylamine (TMA) lyase-catalyzed TMA production from choline (IC50 = 0.9 nM/P. mirabilis & 1.4 nM/D. alaskensis rec. CutC/D) without affecting the bacterial growth (P. mirabilis, E. fergusonii, and P. penneri). A single FMC oral dose (100 mg/kg) to mice on choline-supplemented diet significantly reduces plasma trimethylamine N-oxide (TMAO) level (up to 3 days) as well as diet-induced platelet responsiveness and thrombus formation without toxicity or increased bleeding risk.
Orally active gut bacterial choline trimethylamine (TMA) lyase cutC inhibtor with TMA- & TMA N-oxide (TMAO)-lowering efficacy in vitro and in vivo.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library