Skip to Content
MilliporeSigma

SML3470

MSC-4381

≥98% (HPLC)

Synonym(s):

5-[2-[5-Chloro-2-[[(5-ethoxy-8-quinolinyl)sulfonyl]amino]phenyl]ethynyl]-4-methoxy-2-pyridinecarboxylic acid, MCT4 probe MSC-4381

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H20ClN3O6S
CAS Number:
Molecular Weight:
537.97
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
eCl@ss:
32160703
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=NC=C(C(OC)=C1)C#CC2=CC(Cl)=CC=C2NS(=O)(C3=CC=C(C4=CC=CN=C43)OCC)=O)O

InChI key

FNBUZGRPJLEIMO-UHFFFAOYSA-N

Biochem/physiol Actions

Cell penetrant highly potent and selective inhibitor of monocarboxylate transporter 4 (MCT4)
MSC-4381 is a cell penetrant highly potent and selective inhibitor of monocarboxylate transporter 4 (MCT4). MSC-4381 potently inhibit lactate efflux and intracellular lactate accumulation in MCT4-expressing cell lines.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library