Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3467

Sigma-Aldrich

Remimazolam

≥98% (HPLC)

Synonym(s):

(S)-Methyl 3-(8-bromo-1-methyl-6-(pyridin-2-yl)-4H-benzo[f]imidazo[1,2-a][1,4]diazepin-4-yl)propanoate, CNS 7054 methyl ester, CNS 7056, CNS-7054 methyl ester, CNS-7056, CNS7054 methyl ester, CNS7056, Methyl 3-{(4S)-8-bromo-1-methyl-6-(pyridin-2-yl)-4H-imidazo[1,2-a][1,4]benzodiazepin-4-yl}propanoate, ONO 2745, ONO-2745, ONO2745

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19BrN4O2
CAS Number:
Molecular Weight:
439.31
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule IV

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC1=CN=C2[C@H](CCC(OC)=O)N=C(C3=CC=CC=N3)C4=CC(Br)=CC=C4N12

InChI

1S/C21H19BrN4O2/c1-13-12-24-21-17(7-9-19(27)28-2)25-20(16-5-3-4-10-23-16)15-11-14(22)6-8-18(15)26(13)21/h3-6,8,10-12,17H,7,9H2,1-2H3/t17-/m0/s1

InChI key

CYHWMBVXXDIZNZ-KRWDZBQOSA-N

Biochem/physiol Actions

GABAA receptor positive allosteric modulator (PAM) that causes fast onset and short duration of sedative action in vivo with rapid recovery.
Remimazolam (CNS 7056) is a benzodiazepine derivative with positive allosteric modulator (PAM) activity toward γ-aminobutyric acid type A (GABAA) receptor (pKi = 7.53/7.50/7.56 against 2.5 nM flunitrazepam for binding human/rat/yucatan micropig brain homogenates; pEC50/Emax = 6.44/375% (α1β2γ2), 6.18/186% (α2β2γ2), 6.04/210% (α3β2γ2), 5.86/289% (α5β2γ2s) by whole cell patch clamp using cells expressing respective rat αβγ complex). Intravenous injection (25 mg/kg rats, 15-30 mg/kg mice) causes fast onset and short duration of sedative action with rapid recovery when compared with midazolam.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CNS 7056: a novel ultra-short-acting Benzodiazepine
Anesthesiology, 107, 60-66 (2007)
Metabolism of remimazolam in primary human hepatocytes during continuous long-term infusion in a 3-D bioreactor system
Freyer N, Knospel F, Damm G, Greuel S, Schneider C, Seehofer D, Stohr T, Petersen KU, Zeilinger K
Drug design, development and therapy, 13, 1033-1047 (2019)
Design, Synthesis, and Biological Evaluation of Novel CNS 7056 Derivatives as Sedatives in Rats and Rabbits
Yan Liu , Xiangqing Xu , Jianyong Xie , Huan Ma , Tao Wang , Guisen Zhang , Qingeng Li
Chemical Biology & Drug Design, 88, 38-42 (2016)
Pharmacokinetics and pharmacodynamics of the short-acting sedative CNS 7056 in sheep
R N Upton , A A Somogyi, A M Martinez, J Colvill, C Grant
British Journal of Anaesthesia, 105, 798-809 (2010)
Inhaled Remimazolam Potentiates Inhaled Remifentanil in Rodents
Bevans T, Deering-Rice C, Stockmann C, Rower J, Sakata D, Reilly C
Anesthesia and Analgesia, 124, 1484-1490 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service