Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3446

Sigma-Aldrich

VT107

≥98% (HPLC)

Synonym(s):

(S)-N-(l-(6-Aminopyridin-2-yl)ethyl)-5-(4-(trifluoromethyl)phenyl)-2-naphthamide, N-[(1S)-1-(6-Amino-2-pyridinyl)ethyl]-5-[4-(trifluoromethyl)phenyl]-2-naphthalenecarboxamide, VT 107

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20F3N3O
CAS Number:
Molecular Weight:
435.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

VT107 is a potent pan-TEAD auto-palmitoylation inhibitor (3 μM; using TEAD isotype-overexpressing HEK293T) that blocks TEAD-mediated gene transcription (IC50 = 4.93 nM) by preventing its association with YAP/TAZ. VT107 selectively inhibits the proliferation of mesothelioma cultures with NF2 mutation (NCI-H2051 IC50 = 20.4 nM), deletion (NCI-H2373 IC50 = 9.35 nM) or deficiency (NCI-H226 IC50 = 8.67 nM), but not those with wild-type NF2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manon Sturbaut et al.
European journal of medicinal chemistry, 226, 113835-113835 (2021-09-13)
The Hippo pathway is involved in organ size control and tissue homeostasis by regulating cell growth, proliferation and apoptosis. It controls the phosphorylation of the transcription co-activator YAP (Yes associated protein) and TAZ (Transcriptional coactivator with PDZ-binding motif) in order
Tracy T Tang et al.
Molecular cancer therapeutics, 20(6), 986-998 (2021-04-15)
Mutations in the neurofibromatosis type 2 (NF2) gene that limit or abrogate expression of functional Merlin are common in malignant mesothelioma. Merlin activates the Hippo pathway to suppress nuclear translocation of YAP and TAZ, the major effectors of the pathway

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service