Skip to Content
MilliporeSigma

SML3424

DIM-C-pPhOH

≥98% (HPLC)

Synonym(s):

1,1-Bis(3′-indolyl)-1-(p-hydroxyphenyl)methane, 4-(Di-1H-indol-3-ylmethyl)phenol; (4-Hydroxyphenyl)bis(1H-indol-3-yl)methane, 7,7-Bis(3-indolyl)-p-cresol, C-DIM 8, CDIM8

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H18N2O
CAS Number:
Molecular Weight:
338.40
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

off-white to pink

solubility

DMSO: 2 mg/mL

storage temp.

−20°C

SMILES string

OC1=CC=C(C(C2=CNC3=C2C=CC=C3)C4=CNC5=C4C=CC=C5)C=C1

InChI

1S/C23H18N2O/c26-16-11-9-15(10-12-16)23(19-13-24-21-7-3-1-5-17(19)21)20-14-25-22-8-4-2-6-18(20)22/h1-14,23-26H

InChI key

QCPDFNWJBQMXLI-UHFFFAOYSA-N

Biochem/physiol Actions

DIM-C-pPhOH is a NR4A1 (Nur77) antagonist that exhibit potent anticancer activity in pancreatic, colon, breast, kidney, and rhabdomyosarcoma cells. DIM-C-pPhOH decreases expression of PAX3-FOXO1A and β1-integrin proteins in Rh30 rhabdomyosarcoma and MDA-MB-231breast cancer cells. DIM-C-pPhOH inhibits migration of Rh30 and MDA-MB-231 cancer cells.
NR4A1 (Nur77) antagonist that exhibit potent anticancer activity


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library