Skip to Content
MilliporeSigma

SML3373

BZ194

≥98% (HPLC)

Synonym(s):

3-Carboxy-1-[2-(octylamino)-2-oxoethyl]pyridinium bromide, 3-Carboxy-1-octylcarbamoylmethyl-pyridinium bromide, BZ 194, BZ-194

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H25BrN2O3
CAS Number:
Molecular Weight:
373.29
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C[N+]1=CC(C(O)=O)=CC=C1)NCCCCCCCC.[Br-]

InChI

1S/C16H24N2O3.BrH/c1-2-3-4-5-6-7-10-17-15(19)13-18-11-8-9-14(12-18)16(20)21;/h8-9,11-12H,2-7,10,13H2,1H3,(H-,17,19,20,21);1H

InChI key

LCFXDNPKEWPCGN-UHFFFAOYSA-N

Biochem/physiol Actions

BZ194 is a selective nicotinic acid-adenine dinucleotide phosphate (NAADP) antagonist that blocks NAADP-stimulated ryanodine binding to type 1 ryanodine receptor (RYR-1; IC50 ~10 uM against 1 uM NAADP). BZ194 inhibits Ca(2+) mobilization induced by NAADP (IC50 ~3 uM co-injected with 100 nM NAADP into Jurkat T cells), but not Ins(1,4,5)P3 or cyclic ADP-ribose (cADPR), and exhibits no inhibitory potency against capacitative Ca(2+) entry.
Selective nicotinic acid-adenine dinucleotide phosphate (NAADP) antagonist that blocks NAADP-stimulated ryanodine binding to RYR-1 and calcium mobilization.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library