Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C17H22N2O3S
CAS Number:
Molecular Weight:
334.43
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
COC1=CC=CC(C2=NC=C(S2)CN3CCC(O)(CC3)CO)=C1
InChI key
FXCZNKJONOHACS-UHFFFAOYSA-N
Biochem/physiol Actions
Cell penetrant, potent and low toxic inhibitor of Cryptococcus neoformans sterylglucosidase 1 (Sgl1)
Sgl1 inhibitor Hit 1 is a cell penetrant, potent and low toxic inhibitor of Cryptococcus neoformans sterylglucosidase 1 (Sgl1) that significantly increases in the intracellular concentration of ergosterol 3β-D-glucoside (erg-glc). Sgl1 inhibitor Hit 1 potently inhibits growth of wild-type C. neoformans. Also, it inhibits brain dissemination of C. neoformans in a mouse model of infection.
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.