Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML3334

Sigma-Aldrich

Ceefourin 1

≥95% (HPLC)

Synonym(s):

5-[(2-Benzothiazolylthio)methyl]-2,4-dihydro-4-methyl-3H-1,2,4-triazole-3-thione, Ceefourin-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10N4S3
CAS Number:
Molecular Weight:
294.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CN1C(CSC2=NC3=C(C=CC=C3)S2)=NNC1=S

Biochem/physiol Actions

Ceefourin 1 is a potent and highly selective MRP4 (Multidrug resistance protein 4) inhibitor that exhibits low cellular toxicity and high microsomal and acid stability. Ceefourin 1 inhibits PDAC (pancreatic ductal adenocarcinoma) cell proliferation through MRP4-dependent cAMP extrusion thereby activation of the cAMP/Epac/Rap1 signaling pathway.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service