Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3174

Sigma-Aldrich

Icariside II

≥98% (HPLC)

Synonym(s):

5,7-dihydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-enyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxychromen-4-one, Baohuoside-I, Icariin II, Baohuoside I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O10
CAS Number:
Molecular Weight:
514.52
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -101 to -129°, c = 0.5 in ethanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@@H]([C@H]([C@@H]1C)O)O)O)Oc2[c](c3c([o]c2c4ccc(cc4)OC)c(c(cc3O)O)CC=C(C)C)=O

InChI

1S/C27H30O10/c1-12(2)5-10-16-17(28)11-18(29)19-21(31)26(37-27-23(33)22(32)20(30)13(3)35-27)24(36-25(16)19)14-6-8-15(34-4)9-7-14/h5-9,11,13,20,22-23,27-30,32-33H,10H2,1-4H3/t13-,20-,22+,23+,27-/m0/s1

InChI key

NGMYNFJANBHLKA-LVKFHIPRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Icariside II (Baohuoside-I) is a flavonoid with cytoprotective, neuroprotective, antiproliferative, and erectile function properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service