Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C30H33N5O5S
CAS Number:
Molecular Weight:
575.68
NACRES:
NA.28
UNSPSC Code:
12352107
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
Mpro inhibitor 5h is a potent and selective tight-binding reversible inhibitor of SARS-CoV-2 Mpro that blocks virus replication. It protects VeroE6 cells form SARS-CoV-2WK-521 infection. Mpro inhibitor 5h works synergistically with remdesivir against SARS-CoV-2. Mpro inhibitor 5h shows no significant cytotoxicity to several cell lines at 200 μM.
potent and selective tight-binding reversible inhibitor of SARS-CoV-2 Mpro that blocks virus replication
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.