Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3091

Sigma-Aldrich

Irresistin-16

≥98% (HPLC)

Synonym(s):

7H-Pyrrolo[3,2-f]quinazoline-1,3-diamine,7-([1,1?-biphenyl]-4-ylmethyl), IRS-16

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H19N5
CAS Number:
Molecular Weight:
365.43
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=NC(N)=NC2=C1C3=C(C=C2)N(CC4=CC=C(C=C4)C5=CC=CC=C5)C=C3

InChI

1S/C23H19N5/c24-22-21-18-12-13-28(20(18)11-10-19(21)26-23(25)27-22)14-15-6-8-17(9-7-15)16-4-2-1-3-5-16/h1-13H,14H2,(H4,24,25,26,27)

InChI key

CKQXMSDRDDHCOR-UHFFFAOYSA-N

Biochem/physiol Actions

Irresistin-16, an SCH 79797 derivative, is a broad spectrum bactericidal antibiotic that is effective in both gram-positive and gram-negative bacteria and exhibits low frequency of resistance. Irresistin-16 act through interruption of bacterial folate metabolism and bacterial membrane integrity. It inhibits E. coli dihydrofolate reductase FolA. Irresistin-16 is effective in mouse gonorrhea infection model.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James K Martin et al.
Cell, 181(7), 1518-1532 (2020-06-05)
The rise of antibiotic resistance and declining discovery of new antibiotics has created a global health crisis. Of particular concern, no new antibiotic classes have been approved for treating Gram-negative pathogens in decades. Here, we characterize a compound, SCH-79797, that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service