Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3089

Sigma-Aldrich

MRS-1706

≥98% (HPLC)

Synonym(s):

8-[4-[((4-Acetylphenyl)carbamoylmethyl)oxy]phenyl]-1,3-di(n-propyl)xanthine, MRS 1706, MRS1706, N-(4-Acetylphenyl)-2-(4-(2,6-dioxo-1,3-dipropyl-2,3,6,9-tetrahydro-1H-purin-8-yl)phenoxy)acetamide, N-(4-Acetylphenyl)-2-[4-(2,3,6,9-tetrahydro-2,6-dioxo-1,3-dipropyl-1H-purin-8-yl)phenoxy]acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29N5O5
CAS Number:
Molecular Weight:
503.55
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(COC1=CC=C(C=C1)C2=NC3=C(N2)N(C(N(C3=O)CCC)=O)CCC)NC4=CC=C(C=C4)C(C)=O

InChI

1S/C27H29N5O5/c1-4-14-31-25-23(26(35)32(15-5-2)27(31)36)29-24(30-25)19-8-12-21(13-9-19)37-16-22(34)28-20-10-6-18(7-11-20)17(3)33/h6-13H,4-5,14-16H2,1-3H3,(H,28,34)(H,29,30)

InChI key

ZKUCFFYOQOJLGT-UHFFFAOYSA-N

Biochem/physiol Actions

MRS-1706 is a high-affinity, potent and selective adenosine 2B receptor (A2B) antagonist/inverse agonist (human A1/A2A/A2B/A3 Ki = 157/112/1.39/230 nM; rat A1/A2A Ki = 37.6/548 nM). MRS-1706 (20 nM), but not the A2A antagonist SCH 58261 (20 nM), completely blocks the enhancement of electrically evoked tritium overflow from isolated rat tail artery by 10 μM adenosine receptor agonist NECA in the presence of 30 nM A1 antagonist DPCPX.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Effect of Adenosine A2A and A2B Antagonists on Tracheal Responsiveness, Serum Levels of Cytokines and Lung Inflammation in Guinea Pig Model of Asthma.
Pejman, et al.
Advanced Pharmaceutical Bulletin, 4, 131-138 (2020)
Carmen Diniz et al.
European journal of pharmacology, 504(1-2), 17-25 (2004-10-28)
Adenosine receptors involved in the modulation of noradrenaline release from postganglionic sympathetic nerves in rat tail artery were characterized by studying the effects of adenosine-receptor agonists and antagonists on electrically evoked tritium overflow (100 pulses, 5 Hz) and by immunohistochemistry.
The A2B Adenosine Receptor Modulates the Epithelial- Mesenchymal Transition through the Balance of cAMP/PKA and MAPK/ERK Pathway Activation in Human Epithelial Lung Cells.
Giacomelli, et al.
Frontiers in Pharmacology, 9, 54-54 (2020)
Qilan Li et al.
The Journal of pharmacology and experimental therapeutics, 320(2), 637-645 (2006-11-02)
The human adenosine A(2B) receptor belongs to class A G protein-coupled receptors (GPCRs). In our previous work, constitutively active mutant (CAM) human adenosine A(2B) receptors were identified from a random mutation bank. In the current study, three known A(2B) receptor
Fabio Morandi et al.
Journal of immunology (Baltimore, Md. : 1950), 202(3), 724-735 (2018-12-28)
This study investigates the mechanism(s) underlying the immunoregulatory activities of placenta-derived human amnion epithelial cells (hAEC). The working hypothesis is that NAD+ and ATP, along with ectoenzymes involved in their metabolism, play a significant role in hAEC-mediated immune regulation. Proof

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service