Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2956

Sigma-Aldrich

Nalmefene Hydrochloride

≥98% (HPLC)

Synonym(s):

(5a)-17-(Cyclopropylmethyl)-4,5-epoxy-6-methylene-morphinan-3,14-diol hydrochloride, NIH 1036, Nalmefene HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25NO3·HCl
CAS Number:
Molecular Weight:
375.89
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

C=C1CC[C@@]2(O)[C@H]3CC4=C5[C@@]2(CCN3CC6CC6)[C@H]1OC5=C(C=C4)O.Cl

InChI

1S/C21H25NO3.ClH/c1-12-6-7-21(24)16-10-14-4-5-15(23)18-17(14)20(21,19(12)25-18)8-9-22(16)11-13-2-3-13;/h4-5,13,16,19,23-24H,1-3,6-11H2;1H/t16-,19+,20+,21-;/m1./s1

InChI key

GYWMRGWFQPSQLK-OPHZJPRHSA-N

Biochem/physiol Actions

Nalmefene hydrochloride, a naltrexone analogue, is an inverse agonist at the μ-opioid receptor (MOR) and as a weak partial agonist at the κ-opioid receptor. Nalmefene hydrochloride is used as treatment for addiction.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernadett I Gál et al.
Frontiers in pharmacology, 10, 1087-1087 (2019-10-16)
Nalmefene is approved for as-needed pharmacological treatment in alcohol use disorder (AUD) by the European Medicines Agency. While the cellular effects of nalmefene have been thoroughly investigated, data are very limited on how this agent influences neural signals associated with
Karl Mann et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 26(12), 1941-1949 (2016-11-16)
Nalmefene, a mu- and delta-opioid receptor (MOR, DOR) antagonist and a partial kappa-opioid receptor (KOR) agonist, is approved in the European Union and other countries for the reduction of alcohol consumption in alcohol dependent patients with a high drinking risk
Javier Calleja-Conde et al.
British journal of pharmacology, 173(16), 2490-2505 (2016-05-31)
The opioid antagonist nalmefene (selincro®) was approved for alcohol-related disorders by the European Medicines Agency in 2013. However, there have been no studies regarding the effectiveness of nalmefene when alcohol is used in combination with cocaine. Using operant alcohol self-administration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service