Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2934

Sigma-Aldrich

VZ185

≥98% (HPLC)

Synonym(s):

(2S,4R)-N-(2-(5-(4-(2,6-Dimethoxy-4-(2-methyl-1-oxo-1,2-dihydro-2,7-naphthyridin-4-yl)benzyl)piperazin-1-yl)pentyloxy)-4-(4-methylthiazol-5-yl)benzyl)-1-((S)-2-(1-fluorocyclopropanecarboxamido)-3,3-dimethylbutanoyl)-4-hydroxypyrrolidine-2-carboxamide, VZ 185, VZ-185

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H67FN8O8S
CAS Number:
Molecular Weight:
995.21
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

VZ185 is a potent and selective bromodomain proteins BRD7 and BRD9 degrader that targets BRD7/9 for von Hippel-Lindau (VHL) ubiquitin ligase complex-mediated degradation. VZ185 effectively induces cellular BRD7/9 degradation (by 90% BRD7 & 86% BRD9 in 4 h in HeLa cells, by 84% BRD7 in 2 h in RI-1 cells; BRD7/9 IC50 = 4.5/1.76 nM in 8 h in RI-1 cells) and exerts potent cytotoxicity in BRD9-dependent cancer cultures (IC50 = 3.389 nM/EOL-1 & 39.81 nM/A-204 post 7-day treatment).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vittoria Zoppi et al.
Journal of medicinal chemistry, 62(2), 699-726 (2018-12-13)
Developing PROTACs to redirect the ubiquitination activity of E3 ligases and potently degrade a target protein within cells can be a lengthy and unpredictable process, and it remains unclear whether any combination of E3 and target might be productive for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service