Skip to Content
MilliporeSigma

SML2807

JP4-039

≥98% (HPLC)

Synonym(s):

4-[[(3E,5S)-5-[[(1,1-Dimethylethoxy)carbonyl]amino]-7-methyl-1-oxo-3-octen-1-yl]amino]-2,2,6,6-tetramethyl-1-piperidinyloxy, JP 4-039

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H42N3O4
CAS Number:
Molecular Weight:
424.60
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

faint yellow to dark orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(C/C=C/[C@@H](NC(OC(C)(C)C)=O)CC(C)C)NC1CC(C)(C)N([O])C(C)(C)C1

InChI

1S/C23H42N3O4/c1-16(2)13-17(25-20(28)30-21(3,4)5)11-10-12-19(27)24-18-14-22(6,7)26(29)23(8,9)15-18/h10-11,16-18H,12-15H2,1-9H3,(H,24,27)(H,25,28)/b11-10+/t17-/m1/s1

InChI key

AJHRJWQXDNEJAG-SXSDINLZSA-N

Biochem/physiol Actions

JP4-039 is an electron-/reactive oxygen species (ROS)-scavenging GS-nitroxide composed of 4-amino-TEMPO and a truncated gramicidin S (GS) mitochrondria-targeting sequence. JP4-039 is a superior irradiation mitigator than XJB-5-131 (33% vs. 20% mice survial rate on day 35 with respective compound; 23.6 μmol/kg iv. 24 hr post 9.5 Gy whole body irradiation), while displaying weaker anti-ferroptotic potency (EC50 = 3.58 μM/1.27 μM against 10 μM erastin-/2 μM RSL3-induced HT-1080 ferroptosis vs. 114 nM/68 nM with XJB-5-131) due to lower lipophilicity & mitochondria enrichment. Typical dosing range: 40 nM-10 μM in cultures and 10-20 mg/kg in mice (im., ip., iv.) in vivo.
Mitochondrial-targeted reactive oxygen species (ROS)-scavenging gramicidin S (GS)-nitroxide with in vitro and in vivo efficacy against irradiation-induced damage.


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library