Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML2800

Sigma-Aldrich

Proscillaridin A

≥90% (HPLC)

Synonym(s):

Proscillaridin A, (3β)-3-[(6-Deoxy-α-L-mannopyranosyl)oxy]-14-hydroxybufa-4,20,22-trienolide, 3-(6-Deoxy-α-L-mannopyranosyloxyl)-14-hydroxybufa-4,20,22-trienolide, Scillarenin 3β-rhamnoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H42O8
CAS Number:
Molecular Weight:
530.65
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

C[C@]12CC[C@]3([H])[C@@]4(C)CC[C@H](O[C@]5([H])O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)C=C4CC[C@@]3([H])[C@@]1(O)CC[C@@H]2C6=COC(C=C6)=O

InChI

1S/C30H42O8/c1-16-24(32)25(33)26(34)27(37-16)38-19-8-11-28(2)18(14-19)5-6-22-21(28)9-12-29(3)20(10-13-30(22,29)35)17-4-7-23(31)36-15-17/h4,7,14-16,19-22,24-27,32-35H,5-6,8-13H2,1-3H3/t16-,19?,20?,21?,22?,24-,25+,26+,27-,28?,29?,30?/m0/s1

InChI key

MYEJFUXQJGHEQK-RLQJMXQZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Proscillaridin A, a cardiac glycoside, is an antiarrhythmic drug that enhances DNMT inhibitors and HDAC inhibitors activity. Combination of decitabine with proscillaridin A produced profound gene expression reprogramming associated with down-regulation of 153 epigenetic regulators including SYMD3 and KDM8. Proscillaridin A was recently identified as a potent anti-HBV agent (IC50 = 7.2?nM).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noël J-M Raynal et al.
Molecular cancer therapeutics, 16(2), 397-407 (2016-12-17)
Epigenetic drugs, such as DNA methylation inhibitors (DNMTi) or histone deacetylase inhibitors (HDACi), are approved in monotherapy for cancer treatment. These drugs reprogram gene expression profiles, reactivate tumor suppressor genes (TSG) producing cancer cell differentiation and apoptosis. Epigenetic drugs have
Amara Maryam et al.
Oxidative medicine and cellular longevity, 2018, 3853409-3853409 (2018-03-27)
Cardiac glycosides are natural compounds used for the treatment of cardiovascular disorders. Although originally prescribed for cardiovascular diseases, more recently, they have been rediscovered for their potential use in the treatment of cancer. Proscillaridin A (PSD-A), a cardiac glycoside component
Kaori Okuyama-Dobashi et al.
Scientific reports, 5, 17047-17047 (2015-11-26)
Sodium taurocholate cotransporting polypeptide (NTCP) has been reported as a functional receptor for hepatitis B virus (HBV) infection. However, HBV could not efficiently infect HepG2 cells expressing NTCP (NTCP-HepG2 cells) under adherent monolayer-cell conditions. In this study, NTCP was mainly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service