Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2767

Sigma-Aldrich

HMR-1098

≥98% (HPLC)

Synonym(s):

5-chloro-2-methoxy-N-[2-[4-methoxy-3-[[[(methylamino)thioxomethyl]amino]sulfonyl]phenyl]ethyl]-benzamide sodium salt, HMR 1098

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21ClN3O5S2·Na
CAS Number:
Molecular Weight:
493.96
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[Na]N(CCc2cc(c(cc2)OC)[S](=O)(=O)NC(=S)NC)C(=O)c1c(ccc(c1)Cl)OC

InChI

1S/C19H22ClN3O5S2.Na/c1-21-19(29)23-30(25,26)17-10-12(4-6-16(17)28-3)8-9-22-18(24)14-11-13(20)5-7-15(14)27-2;/h4-7,10-11H,8-9H2,1-3H3,(H3,21,22,23,24,29);/q;+1/p-1

InChI key

SUEVHDKFEXAKAF-UHFFFAOYSA-M

Biochem/physiol Actions

HMR-1098 is a potent and selective blocker of the sarcolemmal ATP-sensitive K+ (sarcKATP) channel. HMR 1098 is inhibits Kir6.2/SUR1-composed KATP channels.
potent and selective blocker of the sarcolemmal ATP-sensitive K+ (sarcKATP) channel

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohui Zhang et al.
International journal of molecular medicine, 38(3), 758-766 (2016-07-20)
The sarcolemmal ATP-sensitive K+ (sarcKATP) channel plays a cardioprotective role during stress. However, the role of the sarcKATP channel in the apoptosis of cardiomyocytes and association with mitochondrial calcium remains unclear. For this purpose, we developed a model of LPS-induced sepsis
Sergey Y Tsibulnikov et al.
General physiology and biophysics, 37(5), 537-547 (2018-10-12)
It was established that adaptation to chronic continuous normobaric hypoxia (CCNH) increases cardiac tolerance to ischemia and reperfusion. It was performed coronary artery occlusion (20 min) and reperfusion (3 h) in Wistar rats. CCNH promoted a decrease in the infarct
Hai Xia Zhang et al.
Journal of molecular and cellular cardiology, 50(3), 552-560 (2010-12-28)
Murine ventricular and atrial ATP-sensitive potassium (K(ATP)) channels contain different sulfonylurea receptors (ventricular K(ATP) channels are Kir6.2/SUR2A complexes, while atrial K(ATP) channels are Kir6.2/SUR1 complexes). HMR 1098, the sodium salt of HMR 1883 {1-[[5-[2-(5-chloro-o-anisamido)ethyl]-2-methoxyphenyl]sulfonyl]-3-methylthiourea}, has been considered as a selective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service