Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2667

Sigma-Aldrich

AC261066

≥98% (HPLC)

Synonym(s):

4-[4-(2-Butoxyethoxy)-5-methyl-2-thiazolyl]-2-fluorobenzoic acid, AC 261066, AC-261066

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20FNO4S
CAS Number:
Molecular Weight:
353.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(ccc(c1)c2[s]c(c(n2)OCCOCCCC)C)C(=O)O

InChI

1S/C17H20FNO4S/c1-3-4-7-22-8-9-23-15-11(2)24-16(19-15)12-5-6-13(17(20)21)14(18)10-12/h5-6,10H,3-4,7-9H2,1-2H3,(H,20,21)

InChI key

HSAOETBFVAWNRP-UHFFFAOYSA-N

Biochem/physiol Actions

AC261066 is an orally active, selective and potent synthetic agonist for the retinoic acid β2-receptor (RARβ2) that decreases oxidative stress in mice fed a high-fat diet. AC261066 reduce ischemia/reperfusion injury of the heart in mice models.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice Marino et al.
The Journal of pharmacology and experimental therapeutics, 366(2), 314-321 (2018-06-17)
We previously discovered that oral treatment with AC261066, a synthetic selective agonist for the retinoic acid β2-receptor, decreases oxidative stress in the liver, pancreas, and kidney of mice fed a high-fat diet (HFD). Since hyperlipidemic states are causally associated with
Steven E Trasino et al.
Journal of molecular medicine (Berlin, Germany), 94(10), 1143-1151 (2016-06-09)
Hepatic stellate cells (HSCs) are an important cellular target for the development of novel pharmacological therapies to prevent and treat nonalcoholic fatty liver diseases (NAFLD). Using a high fat diet (HFD) model of NAFLD, we sought to determine if synthetic
Birgitte W Lund et al.
Journal of medicinal chemistry, 48(24), 7517-7519 (2005-11-24)
4'-Octyl-4-biphenylcarboxylic acid (1g, AC-55649) was identified as a highly isoform-selective agonist at the human RARbeta2 receptor in a functional intact cell-based screening assay. The subsequent hit to lead optimization transformed the lipophilic, poorly soluble hit into a more potent and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service