Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2648

Sigma-Aldrich

Lomibuvir

≥98% (HPLC)

Synonym(s):

5-(3,3-Dimethyl-1-butyn-1-yl)-3-[(trans-4-hydroxycyclohexyl)[(trans-4-methylcyclohexyl)carbonyl]amino]-2-thiophenecarboxylic acid, 5-(3,3-Dimethyl-1-butynyl)-3-[(trans-4-hydroxycyclohexyl)[(trans-4-methylcyclohexyl)carbonyl]amino]thiophene-2-carboxylic acid, 5-(3,3-Dimethyl-but-1-ynyl)-3-[(trans-4-hydroxy-cyclohexyl)-(trans-4-methyl-cyclohexanecarbonyl)-amino]-thiophene-2-carboxylic acid, VCH 222, VCH-222, VCH222, VX 222, VX-222, VX222

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H35NO4S
CAS Number:
Molecular Weight:
445.61
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[s]1c(c(cc1C#CC(C)(C)C)N([C@@H]3CC[C@H](CC3)O)C(=O)[C@@H]2CC[C@H](CC2)C)C(=O)O

InChI

1S/C25H35NO4S/c1-16-5-7-17(8-6-16)23(28)26(18-9-11-19(27)12-10-18)21-15-20(13-14-25(2,3)4)31-22(21)24(29)30/h15-19,27H,5-12H2,1-4H3,(H,29,30)/t16-,17-,18-,19-

InChI key

WPMJNLCLKAKMLA-VVPTUSLJSA-N

Biochem/physiol Actions

Lomibuvir (VX-222; VCH-222) is a non-cytotoxic, orally active non-nucleoside inhibitor (NNI) against hepatitis C virus (HCV) NS5B RNA-dependent RNA polymerase (genotype 1a/1b IC50 = 0.94/1.2 μM). Lomibuvir exhibits HCV genotype-selective antiviral activity in vitro (genotype 1a//1b EC50 = 23.3 nM/12 nM; ineffective against 2a & 2b) and in vivo by targeting HCV NS5B thumb II allosteric pocket, exhibiting no inhibitory potency against human DNA polymerase (α, β, γ IC50 ≥56 μM), respiratory syncytial virus, Influenza A and B, and West Nile virus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nam-Joon Cho et al.
ACS central science, 2(7), 456-466 (2016-08-10)
Therapeutic targeting of membrane-associated viral proteins is complicated by the challenge of investigating their enzymatic activities in the native membrane-bound state. To permit functional characterization of these proteins, we hypothesized that the supported lipid bilayer (SLB) can support in situ
Mohamed Mandour et al.
Antiviral research, 149, 150-153 (2017-11-21)
Combination therapy of directly acting antivirals (DAA's) for the treatment of chronic HCV infections has proven to be a highly effective strategy to cure chronic infections with this virus. Here we studied, using HCV as an example, how to best
Santseharay Ramirez et al.
Hepatology (Baltimore, Md.), 59(2), 395-407 (2013-08-06)
Hepatitis C virus (HCV) is a genetically diverse virus with multiple genotypes exhibiting remarkable differences, particularly in drug susceptibility. Drug and vaccine development will benefit from high-titer HCV cultures mimicking the complete viral life cycle, but such systems only exist
Daryl Humes et al.
Virology, 522, 177-192 (2018-07-23)
The six major epidemiologically important hepatitis C virus (HCV) genotypes differ in global distribution and antiviral responses. Full-length infectious cell-culture adapted clones, the gold standard for HCV studies in vitro, are missing for genotypes 4 and 5. To address this
Johan Winquist et al.
Antiviral research, 97(3), 356-368 (2013-01-12)
Development of allosteric inhibitors into efficient drugs is hampered by their indirect mode-of-action and complex structure-kinetic relationships. To enable the design of efficient allosteric drugs targeting the polymerase of hepatitis C virus (NS5B), the interaction characteristics of three non-nucleoside compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service