Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML2604

Sigma-Aldrich

BAY-885

≥98% (HPLC)

Synonym(s):

BAY 885, BAY885, [2-Amino-4-(trifluoromethoxy)phenyl]{4-[7-(4-methylpiperazin-1-yl)pyrido[3,2-d]pyrimidin-4-yl]piperidin-1-yl}methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28F3N7O2
CAS Number:
Molecular Weight:
515.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N1CCC(CC1)C2=C3C(C=C(C=N3)N4CCN(CC4)C)=NC=N2)C5=CC=C(C=C5N)OC(F)(F)F

Biochem/physiol Actions

BAY-885 is an ATP site-targeting, potent and selective ERK5 (BMK1, MAPK7) inhibitor (IC50 = 35 nM with 250 μM ATP; % inhibition at 1 μM = 62/rat Fer, 58/human EphB3, 43/human EphA5, =20/355 kinases) with no BRD4 affinity (20 μM), nor inhibitory potency toward hERG (10 μM) and CYP enzymes (20 μM; CYP1A2, CYP2C8, CYP2C9, CYP2D6, CYP3A4). BAY-885 potently inhibits 100 ng/mL EGF-induced, ERK5-dependent MEF2 activity (IC50 = 120 nM; SN12C-MEF2-luc reporter assay).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duy Nguyen et al.
Journal of medicinal chemistry, 62(2), 928-940 (2018-12-20)
The availability of a chemical probe to study the role of a specific domain of a protein in a concentration- and time-dependent manner is of high value. Herein, we report the identification of a highly potent and selective ERK5 inhibitor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service