Skip to Content
MilliporeSigma

SML2570

NSC 146109 hydrochloride

≥98% (HPLC)

Synonym(s):

(10-Methyl-9-anthracenyl) methyl carbamimidothioic acid ester hydrochloride, NSC146109 hydrochloride, XI-011 hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H16N2S · HCl
CAS Number:
Molecular Weight:
316.85
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

S(Cc1c2c(c(c3c1cccc3)C)cccc2)\C(=N/[H])\N.Cl

InChI

1S/C17H16N2S.ClH/c1-11-12-6-2-4-8-14(12)16(10-20-17(18)19)15-9-5-3-7-13(11)15;/h2-9H,10H2,1H3,(H3,18,19);1H

InChI key

VIBMUYOXJUCEMA-UHFFFAOYSA-N

Biochem/physiol Actions

NSC146109 is a potent activator of p53 that selectively inhibits growth of transformed cells without genotoxicity. NSC146109 activates p53 by inhibition of MDMX expression.
p53 activator, inhibitor of of MDMX expression


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library