Skip to Content
MilliporeSigma

SML2549

MS645

≥98% (HPLC)

Synonym(s):

(S)-N,N′-(decane-1,10-diyl)bis(2-((6S)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f] [1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)acetamide)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C48H54Cl2N10O2S2
CAS Number:
Molecular Weight:
938.04
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CC1=NN=C2N1C3=C(C(C)=C(C)S3)C(C4=CC=C(Cl)C=C4)=N[C@H]2CC(NCCCCCCCCCCNC(C[C@H]5C6=NN=C(C)N6C7=C(C(C)=C(C)S7)C(C8=CC=C(Cl)C=C8)=N5)=O)=O

InChI

1S/C48H54Cl2N10O2S2/c1-27-29(3)63-47-41(27)43(33-15-19-35(49)20-16-33)53-37(45-57-55-31(5)59(45)47)25-39(61)51-23-13-11-9-7-8-10-12-14-24-52-40(62)26-38-46-58-56-32(6)60(46)48-42(28(2)30(4)64-48)44(54-38)34-17-21-36(50)22-18-34/h15-22,37-38H,7-14,23-26H2,

InChI key

VFGPQYYCEGAFLY-UWXQCODUSA-N

Biochem/physiol Actions

MS645 is a cell penetrant bivalent bromodomain (BRD) inhibitor that inhibits the tandem BD1/BD2 of BRD4. It potently represses BRD4 transcriptional activity in solid-tumor cells including TNBC. MS645 potently inhibits IL-6 expression MDA-MB-231 cells. MS645 exhibits sustained blockade of BRD4 by inhibition of BRD4 interactions with transcription enhancer/mediator proteins MED1 and YY1.
cell penetrant bivalent bromodomain (BRD) inhibitor that inhibits the tandem BD1/BD2 of BRD4


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library