Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2529

Sigma-Aldrich

MDL-800

≥98% (HPLC)

Synonym(s):

Methyl 2-(N-(5-bromo-4-fluoro-2-methylphenyl)sulfamoyl)-5-(3,5-dichlorophenylsulfonamido)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16BrCl2FN2O6S2
CAS Number:
Molecular Weight:
626.30
MDL number:
UNSPSC Code:
41116158
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(cc(c(c1)C)N[S](=O)(=O)c2c(cc(cc2)N[S](=O)(=O)c3cc(cc(c3)Cl)Cl)C(=O)OC)Br

InChI

1S/C21H16BrCl2FN2O6S2/c1-11-5-18(25)17(22)10-19(11)27-35(31,32)20-4-3-14(9-16(20)21(28)33-2)26-34(29,30)15-7-12(23)6-13(24)8-15/h3-10,26-27H,1-2H3

InChI key

FKFQBYBODAKGOA-UHFFFAOYSA-N

Biochem/physiol Actions

MDL-800 is a selective allosteric activator of SIRT6, a member of the SIRT deacetylase family that is responsible for deacetylation of histones H3K9Ac and H3K56ac and has tumor suppressor activity. MDL-800 increased the deacetylase activity of SIRT6 by up to 22-fold resulting in a decrease in H3K9Ac and H3K56Ac in hepatocellular carcinoma (HCC) cells, causing cell cycle arrest and inhibiting their proliferation. It also suppressed the growth of xenografts of the hepatocellular carcinoma cell line BEL7405.
SIRT6 allosteric activator

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhimin Huang et al.
Nature chemical biology, 14(12), 1118-1126 (2018-10-31)
SIRT6, a member of the SIRT deacetylase family, is responsible for deacetylation of histone H3 Nε-acetyl-lysines 9 (H3K9ac) and 56 (H3K56ac). As a tumor suppressor, SIRT6 has frequently been found to have low expression in various cancers. Here, we report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service