Skip to Content
MilliporeSigma

SML2470

GYY4137 sodium, dichloromethane-free

≥98% (HPLC), H2S donor, powder

Synonym(s):

(4-Methoxyphenyl)-4-morpholinyl-phosphinodithioic acid, sodium salt, GYY4137 Na salt, GYY4137 sodium salt, Na-GYY4137, Sodium 4-methoxyphenyl(morpholino)phosphinodithioate, Sodium GYY4137

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H15NNaO2PS2
CAS Number:
Molecular Weight:
311.34
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

GYY4137 sodium, dichloromethane-free, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

[S-]P(C1=CC=C(C=C1)OC)(N2CCOCC2)=S.[Na+]

InChI

1S/C11H16NO2PS2.Na/c1-13-10-2-4-11(5-3-10)15(16,17)12-6-8-14-9-7-12;/h2-5H,6-9H2,1H3,(H,16,17);/q;+1/p-1

InChI key

ZFQXKWOUNSKHAG-UHFFFAOYSA-M

Biochem/physiol Actions

GYY4137 sodium (Na-GYY4137) is a dichloromethane-free, water-soluble sodium salt form of the slow-releasing hydrogen sulfide (H2S) donor GYY4137. Na-GYY4137 is devoid of potential off-target effects associated with the use of dichloromethane-complexed GYY4137 morpholine salt form. Morpholine is biologically active and toxic (LD50 range of 300-500 mg/kg in rodents), while dichloromethane is carcinogenic and can metabolize in vivo to carbon monoxide, causing unintended CO-dependent effects.
Water soluble, non-toxic alternative to the dichloromethane-containing morpholine salt form of the hydrogen sulfide (H2S) donor GYY4137.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library