Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2446

Sigma-Aldrich

TC-A2317 hydrochloride

≥98% (HPLC)

Synonym(s):

2-[(5-Hydroxy-1,5-dimethylhexyl)amino]-4-methyl-6-[(5-methyl-1H-pyrazol-3-yl)amino]-3-pyridinecarbonitrile hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28N6O · HCl
CAS Number:
Molecular Weight:
392.93
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CC1=CC(NC2=NNC(C)=C2)=NC(NC(C)CCCC(C)(C)O)=C1C#N.Cl

InChI

1S/C19H28N6O.ClH/c1-12-9-16(22-17-10-14(3)24-25-17)23-18(15(12)11-20)21-13(2)7-6-8-19(4,5)26;/h9-10,13,26H,6-8H2,1-5H3,(H3,21,22,23,24,25);1H

InChI key

DXVATYHPJJPMIN-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, potent and selective aurora A kinase inhibitor with anti-cancer activity in vitro and in vivo.
TC-A2317 (TC-A 2317) is an orally active, potent and selective aurora A kinase inhibitor (Ki = 1.2 nM; Aurora B Ki = 101 nM; IC50 >1 μM toward 60 other kinases). TC-A2317 inhibits the proliferation of human colorectal carcinoma HCT116 cells in cultures (IC50 = 115 nM) and suppresses HCT116 xenograft-derived tumor growth in mice in vivo (by 59% on day 14; 30 mg/kg/day p.o.) with good pharmacokinectic properties, oral bioavailability (Tmax = 1.2 h, T1/2 = 3.3 h, Cmax = 4930 nM, C60 min = 52 nM in rat serum post 30 mg/kg p.o.), and no adverse effects to the animals. TC-A231 is a racemate with its (S)-enantiomer being more active than the (R)-enantiomer (respective Ki = 0.59 vs. 66 nM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cory T Zumbar et al.
Journal of neuro-oncology, 137(3), 481-492 (2018-02-06)
Glioblastoma is a highly malignant disease in critical need of expanded treatment options. The AURKA inhibitor alisertib exhibits antiproliferative activity against glioblastoma in vitro and in vivo. Unlike current clinically used taxane drugs, the novel taxane TPI 287 penetrates the
Ryoichi Ando et al.
Bioorganic & medicinal chemistry letters, 20(15), 4709-4711 (2010-06-25)
A new class of Aurora A kinase inhibitor was created by transforming 4-(5-methyl-3-pyrazoloamino)pyrimidine moiety of VX-680 to 3-cyano-6-(5-methyl-3pyrazoloamino)pyridine. Compound 6 exhibited a potent Aurora A kinase inhibitory activity, excellent selectivity to Aurora B kinase and other 60 kinases, good cell
Weston Kenneth Ryan et al.
Cell death and differentiation, 26(3), 548-564 (2018-07-28)
The role of mitosis in the progression of precancerous skin remains poorly understood. To address this question, we deleted the mitotic Kinase Aurora-A (Aur-A) in hyperplastic mutant p53 mouse skin as an experimental tool to study the G2/M transition in
Yoo Hong Min et al.
Oncotarget, 7(51), 84718-84735 (2016-10-08)
Mitotic progression is crucial for the maintenance of chromosomal stability. A proper progression is ensured by the activities of multiple kinases. One of these enzymes, the serine/threonine kinase Aurora A, is required for proper mitosis through the regulation of centrosome
Arpan Kumar Rai et al.
Nature, 559(7713), 211-216 (2018-07-06)
Liquid-liquid phase separation has been shown to underlie the formation and disassembly of membraneless organelles in cells, but the cellular mechanisms that control this phenomenon are poorly understood. A prominent example of regulated and reversible segregation of liquid phases may

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service