Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2429

Sigma-Aldrich

3,3′,4-Tri-O-methylellagic acid

≥95% (HPLC)

Synonym(s):

3,3’,4-Tri-O-methylellagic acid, 2,3,8-Tri-O-methylellagic acid, 3-hydroxy-2,7,8-trimethoxy- [1]Benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione, Nasutin B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12O8
CAS Number:
Molecular Weight:
344.27
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

Assay

≥95% (HPLC)

form

solid

storage temp.

−20°C

InChI

1S/C17H12O8/c1-21-9-5-7-11-10-6(16(19)25-15(11)13(9)23-3)4-8(18)12(22-2)14(10)24-17(7)20/h4-5,18H,1-3H3

InChI key

LXEQIOGTMDLLEC-UHFFFAOYSA-N

General description

3,3′,4-Tri-O-methylellagic acid is the methylated analogue of Ellagic acid. 3,3′,4-Tri-O-methylellagic acid was initially isolated from the rhizomes of Euphorbia acaulis.
3,3′,4-Tri-O-methylellagic acid was found to have anti-diabetic properties in mice models treated with Alloxan. Furthermore, 3,3′,4-Tri-O-methylellagic acid was reported to possess anti-bacterial activity against several bacterial strains including, Klebsiella pneumoniae, Enterobacter aerogenes and Staphylococcus aureus.

Other Notes

Solubility: 5mg/ml in DMSO (requires heat), in case of precipitation reheat solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Igor K Voukeng et al.
BMC complementary and alternative medicine, 17(1), 28-28 (2017-01-08)
Elaeophorbia drupifera (Thonn.) Stapf. (Euphorbiaceae) is used in Cameroonian folk medicine to treat several ailments including bacterial-related diseases such as skin infections. In this study, the methanol extract from the leaves (EDL), fractions (EDLa-d), sub-fractions EDLc1-7 and EDLc31-35 as well
Philip F Uzor et al.
EXCLI journal, 15, 290-296 (2016-06-15)
The root of Combretum dolichopetalum (Combreatacea) is used in ethnomedicine for the management of diabetes mellitus. Though some compounds have been isolated from it, the antidiabetic principles have not been identified. The present study was designed to evaluate the chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service