Skip to Content
MilliporeSigma

SML2426

BIBN4096BS

≥95% (HPLC), CGRP receptor antagonist, powder

Synonym(s):

1-[3,5-Dibromo-N-[[4-(1,4-dihydro-2-oxo-3(2H)-quinazolinyl)-1-piperidinyl]carbonyl]-D-tyrosyl-L-lysyl]-4-(4-pyridinyl)-piperazine, 1-[N2-[3,5-Dibromo-N-[[4-(3,4-dihydro-2(1H)-oxoquinazolin-3-yl)-1-piperidinyl]carbonyl]-D-tyrosyl]-L-lysyl]-4-(4-pyridinyl)-piperazine, BIBN 4096, BIBN 4096 BS, Olcegepant, [R-(R*,S*)]-N-[2-[ [5-Amino-1-[[4-(4-pyridinyl)-1-piperazinyl]carbonyl]pentyl]amino]-1-[ (3,5-dibromo-4-hydroxyphenyl)methyl]-2-oxoethyl]-4-(1,4-dihydro-2-oxo-3(2H)-quinazolinyl)-1-piperidinecarboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C38H47Br2N9O5
CAS Number:
Molecular Weight:
869.65
UNSPSC Code:
51111800
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

BIBN4096BS, ≥95% (HPLC)

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

Brc1c(c(cc(c1)CC(NC(=O)N4CCC(CC4)N5Cc6c(cccc6)NC5=O)C(=O)NC(CCCCN)C(=O)N2CCN(CC2)c3ccncc3)Br)O

InChI

1S/C38H47Br2N9O5/c39-29-21-25(22-30(40)34(29)50)23-33(45-37(53)48-15-10-28(11-16-48)49-24-26-5-1-2-6-31(26)44-38(49)54)35(51)43-32(7-3-4-12-41)36(52)47-19-17-46(18-20-47)27-8-13-42-14-9-27/h1-2,5-6,8-9,13-14,21-22,28,32-33,50H,3-4,7,10-12,15-20,23-24,41H2,(H,43,51)(H,44,54)(H,45,53)

InChI key

ITIXDWVDFFXNEG-UHFFFAOYSA-N

Biochem/physiol Actions

BIBN4096BS (Olcegepant) is a potent calcitonin gene-related peptide (CGRP) receptor antagonist with higher affinity/potency toward human over rat & mouse receptor (human/rat Ki = 14.4 pM/3.4 nM by binding assay; human/mouse Kb = 8 pM//7.711 nM by cellular cAMP assay). BIBN4096BS inhibits facial blood flow by electrical stimulation of marmoset monkey trigeminal ganglion (IC50 = 3 μg/kg i.v.) without any intrinsic cardiovascular effects. Studies using anaesthetized rats reveals that BIBN4096BS antagonism against trigeminal nociceptive stimulation by capsaicin is limited to the spinal trigeminal nucleus, but not trigeminal ganglion. Also widely employed for in vivo studies in mice.
Potent calcitonin gene-related peptide (CGRP) receptor antagonist with therapeutic efficacy against migraine in vivo.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library