Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2361

Sigma-Aldrich

Kinesore

>97% (HPLC)

Synonym(s):

3,5-Dibromo-4-hydroxy-benzoic acid 2-[[2,5-dimethyl-1-(3-nitrophenyl)-1H-pyrrol-3-yl]methylene]hydrazide, 3,5-Dibromo-N′-{[2,5-dimethyl-1-(3-nitrophenyl)-1H-pyrrol-3-yl]methylene}-4-hydroxybenzohydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16Br2N4O4
CAS Number:
Molecular Weight:
536.17
MDL number:
UNSPSC Code:
31401503
NACRES:
NA.77

Assay

>97% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Brc1c(c(cc(c1)C(=O)N\N=C\c2c([n](c(c2)C)c3cc(ccc3)[N+](=O)[O-])C)Br)O

InChI

1S/C20H16Br2N4O4/c1-11-6-14(12(2)25(11)15-4-3-5-16(9-15)26(29)30)10-23-24-20(28)13-7-17(21)19(27)18(22)8-13/h3-10,27H,1-2H3,(H,24,28)/b23-10+

InChI key

DUGCMEGLYHBMAR-AUEPDCJTSA-N

Biochem/physiol Actions

Kinesore is a cell penetrant and potent activator of kinesin-1 that induces kinesin light chain tetratricopeptide repeat domain-dependent kinesin-1 activation in the absence of organelle cargo-adaptor engagement. It inhibits kinesin-1 interaction with peptide fragment of organelle-specific cargo adaptors. Kinesore induces kinesin light chain conformational switch that causes remodeling of the microtubule network in cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas S Randall et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(52), 13738-13743 (2017-12-13)
The microtubule motor kinesin-1 interacts via its cargo-binding domain with both microtubules and organelles, and hence plays an important role in controlling organelle transport and microtubule dynamics. In the absence of cargo, kinesin-1 is found in an autoinhibited conformation. The
Yerbol Tagay et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 10(31), e2302229-e2302229 (2023-09-20)
The principal cause of death in cancer patients is metastasis, which remains an unresolved problem. Conventionally, metastatic dissemination is linked to actomyosin-driven cell locomotion. However, the locomotion of cancer cells often does not strictly line up with the measured actomyosin
Raeann Goering et al.
eLife, 12 (2023-03-04)
RNA molecules are localized to specific subcellular regions through interactions between RNA regulatory elements and RNA binding proteins (RBPs). Generally, our knowledge of the mechanistic details behind the localization of a given RNA is restricted to a particular cell type.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service