Skip to Content
MilliporeSigma

SML2357

Isavuconazole

≥98% (HPLC), powder, fungal sterol biosynthesis inhibitor

Synonym(s):

(2R,3R)-3-[4-(4-Cyanophenyl)thiazol-2-yl]-2-(2,5-difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butan-2-ol, 1-[(2R,3R)-3-[4-(4-Cyanophenyl)thiazol-2-yl]-2-(2,5-difluorophenyl)-2-hydroxybutyl]-1,2,4-triazole, 4-[2-[(1R,2R)-2-(2,5-Difluorophenyl)-2-hydroxy-1-methyl-3-(1H-1,2,4-triazol-1-yl)propyl]-4-thiazolyl]benzonitrile, BAL 4815, RO 0094815

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H17F2N5OS
CAS Number:
Molecular Weight:
437.47
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Isavuconazole, ≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(cc(cc1)F)[C@](O)([C@@H](C)c3[s]cc(n3)c4ccc(cc4)C#N)C[n]2ncnc2

InChI

1S/C22H17F2N5OS/c1-14(21-28-20(10-31-21)16-4-2-15(9-25)3-5-16)22(30,11-29-13-26-12-27-29)18-8-17(23)6-7-19(18)24/h2-8,10,12-14,30H,11H2,1H3/t14-,22+/m0/s1

InChI key

DDFOUSQFMYRUQK-RCDICMHDSA-N

Application

Isavuconazole has been used as an antifungal agent to test its effect on Mucor circinelloides gene transcription. It has also been used
to analyze its in vitro activity against isolates from the R. argillacea species complex A. fumigatus, N. hiratsukae and C. parapsilosis.

Biochem/physiol Actions

Isavuconazole is a triazole antifungal drug. It inhibits fungal sterol biosynthesis by inhibition of cytochrome P450 sterol 14-α-demethylase (CYP51), an enzyme in the sterol biosynthesis pathway that leads from lanosterol to ergosterol. It has been approved for the treatment of both invasive aspergillosis and invasive mucormycosis.
Triazole antifungal drug.


Pictograms

Health hazard

Signal Word

Warning

Hazard Codes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library