Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML2351

Sigma-Aldrich

Dabigatran etexilate

≥98% (HPLC)

Synonym(s):

N-[[2-[[[4-[[[(Hexyloxy)carbonyl]amino]iminomethyl] phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-β-alanine ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H41N7O5
CAS Number:
Molecular Weight:
627.73
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OCCCCCC)NC(C(C=C1)=CC=C1NCC2=NC3=CC(C(N(CCC(OCC)=O)C4=NC=CC=C4)=O)=CC=C3N2C)=N

InChI

1S/C34H41N7O5/c1-4-6-7-10-21-46-34(44)39-32(35)24-12-15-26(16-13-24)37-23-30-38-27-22-25(14-17-28(27)40(30)3)33(43)41(20-18-31(42)45-5-2)29-11-8-9-19-36-29/h8-9,11-17,19,22,37H,4-7,10,18,20-21,23H2,1-3H3,(H2,35,39,44)

InChI key

KSGXQBZTULBEEQ-UHFFFAOYSA-N

Biochem/physiol Actions

Dabigatran etexilate is a prodrug of dabigatran, a nonpeptide direct thrombin inhibitor. Dabigatron is an anticoagulant that functions by directly inhibiting both free and fibrin-bound thrombin. Dabigatron has been shown to prevent strokes in those with atrial fibrillation not caused by heart valve issues.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wolfgang G Eisert et al.
Arteriosclerosis, thrombosis, and vascular biology, 30(10), 1885-1889 (2010-07-31)
Dabigatran is a highly selective, reversible, and potent thrombin inhibitor and is orally available as the prodrug, dabigatran etexilate. It has shown antithrombotic efficacy in animal models of thrombosis, with a rapid onset of action and predictable pharmacodynamic response. Peak

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service