Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2348

Sigma-Aldrich

BAY-707

≥98% (HPLC)

Synonym(s):

(S)-N-Ethyl-4-(3-methylmorpholino)-1H-pyrrolo[2,3-b]pyridine-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20N4O2
CAS Number:
Molecular Weight:
288.34
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NCC)C1=CC2=C(N1)N=CC=C2N3CCOC[C@@H]3C

InChI

1S/C15H20N4O2/c1-3-16-15(20)12-8-11-13(4-5-17-14(11)18-12)19-6-7-21-9-10(19)2/h4-5,8,10H,3,6-7,9H2,1-2H3,(H,16,20)(H,17,18)/t10-/m0/s1

InChI key

RPMGXDCRCWWCRY-JTQLQIEISA-N

Biochem/physiol Actions

BAY-707 is an inhibitor of the human 7,8-dihydro-8-oxoguaninetriphosphatase MTH1 (NUDT1), which hydrolyzes oxidized purine nucleoside triphosphates that might otherwise be incorporated into DNA/RNA and contribute to DNA damage. BAY-707 was shown to be a substrate-competitive, selective MTH1 inhibitor with an IC50 value of 2.3 nM.
Inhibitor of the human 7,8-dihydro-8-oxoguaninetriphosphatase MTH1 (NUDT1)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel Ellermann et al.
ACS chemical biology, 12(8), 1986-1992 (2017-07-06)
MTH1 is a hydrolase responsible for sanitization of oxidized purine nucleoside triphosphates to prevent their incorporation into replicating DNA. Early tool compounds published in the literature inhibited the enzymatic activity of MTH1 and subsequently induced cancer cell death; however recent
Fredrik Rahm et al.
Journal of medicinal chemistry, 61(6), 2533-2551 (2018-02-28)
Recent literature has both suggested and questioned MTH1 as a novel cancer target. BAY-707 was just published as a target validation small molecule probe for assessing the effects of pharmacological inhibition of MTH1 on tumor cell survival, both in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service